siRNA Details
Gene Symbol: FRMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244959.1 | NP_001231888.1 | 14 | 1503 | ||
NM_001244960.1 | NP_001231889.1 | 14 | 1279 | |||
NM_001244961.1 | NP_001231890.1 | 9 | 815 | |||
NM_001244962.1 | NP_001231891.1 | 2 | 382 | |||
NM_174938.5 | NP_777598.3 | 14 | 1503 | |||
XM_017014588.1 | XP_016870077.1 | 14 | 1193 | |||
XM_017014589.1 | XP_016870078.1 | 9 | 3630 | |||
XM_017014590.1 | XP_016870079.1 | 13 | 1416 | |||
XM_017014591.1 | XP_016870080.1 | 2 | 394 | |||
GenBank mRNA | AK094281.1 | - | 14 | 1204 | ||
AK223597.1 | - | 14 | 1430 | |||
AK297722.1 | - | 14 | 1421 | |||
AY137774.1 | - | 14 | 1287 | |||
BC023560.1 | - | 2 | 382 | |||
BC037253.1 | - | 9 | 815 | 5166212 | ||
BX647549.1 | - | 1 | 65 | |||
EF560742.1 | - | 14 | 1354 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
