siRNA Details
Gene Symbol: NEFH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021076.3 | NP_066554.2 | 4 | 1796 | ||
GenBank mRNA | AB020652.2 | - | 6 | 1990 | ||
AF203032.1 | - | 4 | 1725 | |||
AK302660.1 | - | 4 | 1796 | |||
BC008648.1 | - | 1 | 73 | |||
BC073969.1 | - | 4 | 1758 | 6137962 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
