siRNA Details
Gene Symbol: SH2D1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114937.2 | NP_001108409.1 | 3 | 645 | ||
NM_002351.4 | NP_002342.1 | 3 | 645 | |||
GenBank mRNA | AB586694.1 | - | 3 | 386 | ||
AF072930.1 | - | 3 | 370 | |||
AF073019.1 | - | 3 | 370 | |||
AF100539.1 | - | 3 | 283 | |||
AF100540.1 | - | 2 | 274 | |||
AF100541.1 | - | 3 | 338 | |||
AF100542.1 | - | 3 | 261 | |||
AF100543.1 | - | 2 | 219 | |||
AK311645.1 | - | 2 | 144 | |||
AK311911.1 | - | 3 | 319 | |||
AL023657.1 | - | 3 | 583 | |||
BC020732.1 | - | 3 | 377 | 4702653 | ||
CR542031.1 | - | 3 | 284 | |||
CR542043.1 | - | 3 | 284 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
