siRNA Details
Gene Symbol: MUC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135091.1 | NP_001128563.1 | 3 | 481 | ||
NM_001135092.1 | NP_001128564.1 | 3 | 481 | |||
NM_145650.3 | NP_663625.2 | 2 | 393 | |||
XM_017017235.1 | XP_016872724.1 | 3 | 481 | |||
XM_017017236.1 | XP_016872725.1 | 3 | 481 | |||
GenBank mRNA | AJ417818.1 | - | 3 | 349 | ||
AJ507429.1 | - | 3 | 355 | |||
AK128337.1 | - | 2 | 246 | |||
AY358668.1 | - | 3 | 446 | |||
BC020912.1 | - | 3 | 473 | 4733074 | ||
BC058007.1 | - | 3 | 359 | 4719841 | ||
DA842986.1 | - | 3 | 482 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
