siRNA Details
Gene Symbol: ANKRD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256182.1 | NP_001243111.1 | 10 | 2498 | ||
NM_001256183.1 | NP_001243112.1 | 9 | 2424 | |||
NM_013275.5 | NP_037407.4 | 9 | 2427 | |||
XM_006721181.1 | XP_006721244.1 | 9 | 2204 | |||
XM_006721184.2 | XP_006721247.1 | 8 | 2242 | |||
XM_011523051.2 | XP_011521353.1 | 10 | 2487 | |||
XM_011523053.2 | XP_011521355.1 | 10 | 2593 | |||
XM_011523054.2 | XP_011521356.1 | 10 | 2507 | |||
XM_011523055.2 | XP_011521357.1 | 11 | 2792 | |||
XM_011523056.2 | XP_011521358.1 | 10 | 2529 | |||
XM_011523057.2 | XP_011521359.1 | 9 | 2410 | |||
XM_017023182.1 | XP_016878671.1 | 9 | 2860 | |||
XM_017023183.1 | XP_016878672.1 | 10 | 2596 | |||
XM_017023184.1 | XP_016878673.1 | 10 | 2593 | |||
XM_017023185.1 | XP_016878674.1 | 8 | 2234 | |||
XM_017023186.1 | XP_016878675.1 | 12 | 2970 | |||
XM_017023187.1 | XP_016878676.1 | 8 | 2322 | |||
GenBank mRNA | AK290994.1 | - | 8 | 2106 | ||
AY131227.1 | - | 1 | 823 | |||
AY373756.1 | - | 9 | 2430 | |||
AY533563.1 | - | 10 | 2255 | |||
BC017437.1 | - | 9 | 2396 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
