siRNA Details
Gene Symbol: CLSTN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014718.3 | NP_055533.2 | 17 | 2895 | ||
XM_006719163.3 | XP_006719226.1 | 16 | 4221 | |||
GenBank mRNA | AB018269.1 | - | 16 | 3191 | ||
AJ277460.1 | - | 17 | 2892 | |||
AK299177.1 | - | 17 | 2869 | |||
AK299246.1 | - | 8 | 1097 | |||
AK316059.1 | - | 17 | 2895 | |||
AY753302.1 | - | 16 | 2653 | |||
BC039075.1 | - | 2 | 1115 | |||
BC104767.1 | - | 17 | 2699 | 8143770 | ||
BC111491.1 | - | 17 | 2883 | 5213830 | ||
BC112283.1 | - | 17 | 2711 | 8327751 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
