siRNA Details
Gene Symbol: LRRC28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284400.2 | NP_001271329.1 | 9 | 1330 | ||
NM_001321675.1 | NP_001308604.1 | 10 | 1486 | |||
NM_001321676.1 | NP_001308605.1 | 11 | 1478 | |||
NM_001321677.1 | NP_001308606.1 | 9 | 1283 | |||
NM_001321678.1 | NP_001308607.1 | 9 | 1178 | |||
NM_001321679.1 | NP_001308608.1 | 9 | 1449 | |||
NM_001321680.1 | NP_001308609.1 | 9 | 1445 | |||
NM_144598.4 | NP_653199.2 | 10 | 1490 | |||
NR_135753.1 | 10 | 1395 | ||||
NR_135754.1 | 10 | 1243 | ||||
NR_135755.1 | 9 | 1350 | ||||
NR_135756.1 | 9 | 1354 | ||||
NR_135757.1 | 11 | 1464 | ||||
NR_135758.1 | 10 | 1518 | ||||
NR_135759.1 | 8 | 1238 | ||||
XM_011521218.2 | XP_011519520.1 | 10 | 1526 | |||
GenBank mRNA | AK091588.1 | - | 9 | 1352 | ||
AK127177.1 | - | 5 | 1173 | |||
AK295161.1 | - | 9 | 1158 | |||
AK296352.1 | - | 10 | 1297 | |||
AY358082.1 | - | 9 | 1187 | |||
BC019704.1 | - | 4 | 704 | |||
BC047306.1 | - | 10 | 1405 | 4999965 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
