siRNA Details
Gene Symbol: LRRC28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284400.2 | NP_001271329.1 | 9 | 1195 | ||
NM_001321675.1 | NP_001308604.1 | 10 | 1351 | |||
NM_001321676.1 | NP_001308605.1 | 11 | 1343 | |||
NM_001321677.1 | NP_001308606.1 | 9 | 1148 | |||
NM_001321678.1 | NP_001308607.1 | 9 | 1043 | |||
NM_001321679.1 | NP_001308608.1 | 9 | 1314 | |||
NM_001321680.1 | NP_001308609.1 | 9 | 1310 | |||
NM_144598.4 | NP_653199.2 | 10 | 1355 | |||
NR_135753.1 | 10 | 1260 | ||||
NR_135754.1 | 10 | 1108 | ||||
NR_135755.1 | 9 | 1215 | ||||
NR_135756.1 | 9 | 1219 | ||||
NR_135757.1 | 11 | 1329 | ||||
NR_135758.1 | 10 | 1383 | ||||
NR_135759.1 | 8 | 1103 | ||||
XM_011521218.2 | XP_011519520.1 | 10 | 1391 | |||
GenBank mRNA | AK091588.1 | - | 9 | 1217 | ||
AK127177.1 | - | 5 | 1038 | |||
AK292887.1 | - | 10 | 1212 | |||
AK295161.1 | - | 9 | 1023 | |||
AY358082.1 | - | 9 | 1052 | |||
BC019704.1 | - | 4 | 569 | |||
BC047306.1 | - | 10 | 1270 | 4999965 | ||
DA787321.1 | - | 3 | 492 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
