siRNA Details
Gene Symbol: LRRC28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284400.2 | NP_001271329.1 | 9 | 1248 | ||
NM_001321675.1 | NP_001308604.1 | 10 | 1404 | |||
NM_001321676.1 | NP_001308605.1 | 11 | 1396 | |||
NM_001321677.1 | NP_001308606.1 | 9 | 1201 | |||
NM_001321678.1 | NP_001308607.1 | 9 | 1096 | |||
NM_001321679.1 | NP_001308608.1 | 9 | 1367 | |||
NM_001321680.1 | NP_001308609.1 | 9 | 1363 | |||
NM_144598.4 | NP_653199.2 | 10 | 1408 | |||
NR_135753.1 | 10 | 1313 | ||||
NR_135754.1 | 10 | 1161 | ||||
NR_135755.1 | 9 | 1268 | ||||
NR_135756.1 | 9 | 1272 | ||||
NR_135757.1 | 11 | 1382 | ||||
NR_135758.1 | 10 | 1436 | ||||
NR_135759.1 | 8 | 1156 | ||||
XM_011521218.2 | XP_011519520.1 | 10 | 1444 | |||
GenBank mRNA | AK091588.1 | - | 9 | 1270 | ||
AK127177.1 | - | 5 | 1091 | |||
AK292887.1 | - | 10 | 1265 | |||
AK295161.1 | - | 9 | 1076 | |||
AK296352.1 | - | 10 | 1215 | |||
AY358082.1 | - | 9 | 1105 | |||
BC019704.1 | - | 4 | 622 | |||
BC047306.1 | - | 10 | 1323 | 4999965 | ||
DA787321.1 | - | 3 | 545 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
