siRNA Details
Gene Symbol: ZNF501 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258280.1 | NP_001245209.1 | 3 | 910 | ||
NM_145044.3 | NP_659481.2 | 3 | 617 | |||
GenBank mRNA | AK054613.1 | - | 3 | 614 | ||
AK297216.1 | - | 3 | 599 | |||
AK316574.1 | - | 3 | 617 | |||
BC013762.1 | - | 3 | 592 | 4521303 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
