siRNA Details
Gene Symbol: CLN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000086.2 | NP_000077.1 | 12 | 1260 | ||
NM_001042432.1 | NP_001035897.1 | 13 | 1296 | |||
NM_001286104.1 | NP_001273033.1 | 12 | 1224 | |||
NM_001286105.1 | NP_001273034.1 | 10 | 1101 | |||
NM_001286109.1 | NP_001273038.1 | 10 | 1109 | |||
NM_001286110.1 | NP_001273039.1 | 11 | 1181 | |||
GenBank mRNA | AF015593.1 | - | 12 | 991 | ||
AF015594.1 | - | 11 | 952 | |||
AF015595.1 | - | 10 | 811 | |||
AF015598.1 | - | 10 | 745 | |||
AF015600.1 | - | 8 | 667 | |||
AF077956.1 | - | 12 | 937 | |||
AF077957.1 | - | 12 | 937 | |||
AF077958.1 | - | 12 | 937 | |||
AF077959.1 | - | 12 | 937 | |||
AF077960.1 | - | 11 | 793 | |||
AF077961.1 | - | 12 | 937 | |||
AF077962.1 | - | 12 | 937 | |||
AF077963.1 | - | 12 | 937 | |||
AF077964.1 | - | 10 | 720 | |||
AF077965.1 | - | 9 | 607 | |||
AF077966.1 | - | 12 | 937 | |||
AF077967.1 | - | 11 | 857 | |||
AF077968.1 | - | 10 | 720 | |||
AF077969.1 | - | 8 | 551 | |||
AF077970.1 | - | 8 | 551 | |||
AF077971.1 | - | 12 | 937 | |||
AF077972.1 | - | 11 | 886 | |||
AF077973.1 | - | 10 | 778 | |||
AF078169.1 | - | 12 | 937 | |||
AK090709.1 | - | 12 | 2465 | |||
AK294070.1 | - | 11 | 1105 | |||
AK297690.1 | - | 10 | 1043 | |||
AK302027.1 | - | 12 | 964 | |||
AK302138.1 | - | 12 | 985 | |||
AK313002.1 | - | 13 | 1068 | |||
BC002394.2 | - | 13 | 1054 | 2820893 | ||
BC004433.1 | - | 13 | 1054 | 2820893 | ||
BC111068.1 | - | 10 | 1039 | 5764535 | ||
EF587244.1 | - | 5 | 314 | |||
U32680.1 | - | 12 | 1074 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
