siRNA Details
Gene Symbol: FAM78A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033387.3 | NP_203745.2 | 2 | 1466 | ||
XM_011518567.1 | XP_011516869.1 | 3 | 1009 | |||
XM_011518568.2 | XP_011516870.1 | 2 | 1712 | |||
GenBank mRNA | AK024434.1 | - | 3 | 2088 | ||
AK095423.1 | - | 2 | 1494 | |||
BC029924.2 | - | 2 | 880 | |||
BC037559.1 | - | 4 | 1187 | |||
BC049388.2 | - | 2 | 1466 | 6050655 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
