siRNA Details
Gene Symbol: TMEM138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016464.4 | NP_057548.1 | 3 | 805 | ||
NR_028473.1 | 4 | 688 | ||||
XM_006718585.3 | XP_006718648.1 | 3 | 445 | |||
XM_006718586.2 | XP_006718649.1 | 3 | 445 | |||
XM_006718588.3 | XP_006718651.1 | 3 | 300 | |||
XM_011545098.2 | XP_011543400.1 | 3 | 445 | |||
XM_011545099.1 | XP_011543401.1 | 3 | 445 | |||
XM_011545100.1 | XP_011543402.1 | 3 | 445 | |||
XM_017017917.1 | XP_016873406.1 | 3 | 445 | |||
GenBank mRNA | AF151030.1 | - | 3 | 422 | ||
AF151032.1 | - | 3 | 419 | |||
AK027724.1 | - | 4 | 408 | |||
AK303219.1 | - | 3 | 420 | |||
AK307371.1 | - | 4 | 319 | |||
AL832879.1 | - | 3 | 655 | |||
BC005201.1 | - | 3 | 420 | 3684390 | ||
CB956757.1 | - | 4 | 360 |
