siRNA Details
Gene Symbol: CYP2R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024514.4 | NP_078790.2 | 3 | 455 | ||
XM_005252788.1 | XP_005252845.1 | 4 | 745 | |||
XM_005252789.2 | XP_005252846.1 | 4 | 998 | |||
XM_011519895.1 | XP_011518197.1 | 4 | 1149 | |||
XM_011519898.2 | XP_011518200.1 | 3 | 6779 | |||
XM_017017190.1 | XP_016872679.1 | 3 | 3340 | |||
XM_017017191.1 | XP_016872680.1 | 3 | 960 | |||
XM_017017192.1 | XP_016872681.1 | 4 | 945 | |||
XM_017017193.1 | XP_016872682.1 | 3 | 907 | |||
XM_017017194.1 | XP_016872683.1 | 3 | 760 | |||
XM_017017195.1 | XP_016872684.1 | 3 | 2503 | |||
GenBank mRNA | AY323817.1 | - | 3 | 455 | ||
BC104907.1 | - | 3 | 500 | 8143910 | ||
BC104909.1 | - | 3 | 502 | 8143912 | ||
BC111070.1 | - | 3 | 740 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
