siRNA Details
Gene Symbol: ANKRD13A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033121.1 | NP_149112.1 | 15 | 2280 | ||
XM_005253980.2 | XP_005254037.1 | 15 | 2323 | |||
XM_005253981.3 | XP_005254038.1 | 15 | 2280 | |||
XM_005253982.3 | XP_005254039.1 | 15 | 2277 | |||
XM_005253984.1 | XP_005254041.1 | 15 | 2079 | |||
XM_011538937.1 | XP_011537239.1 | 14 | 2179 | |||
XM_011538938.2 | XP_011537240.1 | 14 | 2122 | |||
XM_017020157.1 | XP_016875646.1 | 14 | 2169 | |||
XM_017020158.1 | XP_016875647.1 | 14 | 2163 | |||
XM_017020159.1 | XP_016875648.1 | 15 | 2063 | |||
XM_017020160.1 | XP_016875649.1 | 15 | 2063 | |||
XM_017020161.1 | XP_016875650.1 | 15 | 2060 | |||
XM_017020162.1 | XP_016875651.1 | 14 | 2116 | |||
XM_017020163.1 | XP_016875652.1 | 14 | 2277 | |||
GenBank mRNA | AF155103.1 | - | 7 | 1108 | ||
AK022658.1 | - | 6 | 965 | |||
AK025261.1 | - | 4 | 1004 | |||
AK095130.1 | - | 4 | 711 | |||
AK130522.1 | - | 6 | 1689 | |||
AK293232.1 | - | 11 | 1602 | |||
AY364251.1 | - | 15 | 2021 | |||
BC032833.2 | - | 15 | 2121 | 4830174 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
