siRNA Details
Gene Symbol: LRCH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032773.3 | NP_116162.1 | 18 | 2018 | ||
XM_005269362.2 | XP_005269419.1 | 18 | 2389 | |||
XM_005269363.2 | XP_005269420.1 | 17 | 2317 | |||
XM_005269365.2 | XP_005269422.1 | 17 | 2281 | |||
XM_005269367.2 | XP_005269424.1 | 16 | 2209 | |||
XM_006713791.3 | XP_006713854.1 | 18 | 2389 | |||
XM_006713792.3 | XP_006713855.1 | 17 | 2317 | |||
XM_006713793.2 | XP_006713856.1 | 17 | 2281 | |||
XM_017007350.1 | XP_016862839.1 | 18 | 2389 | |||
XM_017007351.1 | XP_016862840.1 | 16 | 2263 | |||
XM_017007352.1 | XP_016862841.1 | 15 | 2155 | |||
GenBank mRNA | AK024647.1 | - | 3 | 224 | ||
AK055819.1 | - | 13 | 1124 | |||
AK293745.1 | - | 18 | 2007 | |||
AK300466.1 | - | 15 | 1518 | |||
AK303521.1 | - | 17 | 1824 | |||
AL137527.1 | - | 1 | 97 | |||
BC007504.2 | - | 18 | 1970 | 2960711 | ||
BC010565.1 | - | 3 | 243 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
