siRNA Details
Gene Symbol: LRCH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032773.3 | NP_116162.1 | 19 | 2092 | ||
XM_005269362.2 | XP_005269419.1 | 19 | 2463 | |||
XM_005269363.2 | XP_005269420.1 | 18 | 2391 | |||
XM_005269365.2 | XP_005269422.1 | 18 | 2355 | |||
XM_005269367.2 | XP_005269424.1 | 17 | 2283 | |||
XM_006713791.3 | XP_006713854.1 | 19 | 2463 | |||
XM_006713792.3 | XP_006713855.1 | 18 | 2391 | |||
XM_006713793.2 | XP_006713856.1 | 18 | 2355 | |||
XM_017007350.1 | XP_016862839.1 | 19 | 2463 | |||
XM_017007351.1 | XP_016862840.1 | 17 | 2337 | |||
XM_017007352.1 | XP_016862841.1 | 16 | 2229 | |||
GenBank mRNA | AK024647.1 | - | 4 | 298 | ||
AK055819.1 | - | 14 | 1198 | |||
AK293745.1 | - | 19 | 2081 | |||
AK300466.1 | - | 16 | 1592 | |||
AK303521.1 | - | 18 | 1898 | |||
AL137527.1 | - | 2 | 171 | |||
BC007504.2 | - | 19 | 2044 | 2960711 | ||
BC010565.1 | - | 4 | 317 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
