siRNA Details
Gene Symbol: CCNK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099402.1 | NP_001092872.1 | 8 | 1092 | ||
XM_005268154.4 | XP_005268211.1 | 8 | 1217 | |||
XM_011537275.2 | XP_011535577.1 | 8 | 1389 | |||
GenBank mRNA | AB209373.1 | - | 3 | 3486 | ||
AK311075.1 | - | 8 | 1070 | |||
BQ432222.1 | - | 5 | 668 | |||
BX247958.1 | - | 1 | 128 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
