siRNA Details
Gene Symbol: CCNK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099402.1 | NP_001092872.1 | 11 | 1747 | ||
XM_005268154.4 | XP_005268211.1 | 11 | 1872 | |||
XM_011537275.2 | XP_011535577.1 | 11 | 2044 | |||
GenBank mRNA | AB209373.1 | - | 5 | 4767 | ||
BC015935.1 | - | 9 | 1053 | 3907416 | ||
BT006950.1 | - | 8 | 944 | |||
BX247958.1 | - | 4 | 783 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
