siRNA Details
Gene Symbol: SCGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006998.3 | NP_008929.2 | 10 | 872 | ||
XM_017010181.1 | XP_016865670.1 | 9 | 584 | |||
GenBank mRNA | AK289477.1 | - | 10 | 927 | ||
BC000336.2 | - | 10 | 839 | 2822613 | ||
BC003036.1 | - | 10 | 839 | 2822613 | ||
Y16752.1 | - | 10 | 825 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
