siRNA Details
Gene Symbol: SCGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006998.3 | NP_008929.2 | 3 | 391 | ||
XM_017010181.1 | XP_016865670.1 | 2 | 103 | |||
GenBank mRNA | AF420280.1 | - | 2 | 117 | ||
AK289477.1 | - | 3 | 446 | |||
BC000336.2 | - | 3 | 358 | 2822613 | ||
BC003036.1 | - | 3 | 358 | 2822613 | ||
BM511122.1 | - | 3 | 397 | |||
Y16752.1 | - | 3 | 344 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
