siRNA Details
Gene Symbol: CASC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026939.1 | 5 | 629 | |||
NR_026940.1 | 4 | 529 | ||||
GenBank mRNA | AJ535621.1 | - | 4 | 342 | ||
AY226450.1 | - | 5 | 445 | |||
BC036393.1 | - | 5 | 630 | |||
BC112306.1 | - | 4 | 353 | |||
BC112308.1 | - | 4 | 353 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
