siRNA Details
Gene Symbol: CASC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026939.1 | 4 | 511 | |||
GenBank mRNA | AY226450.1 | - | 4 | 327 | ||
BC036393.1 | - | 4 | 513 | |||
BC112306.1 | - | 3 | 235 | |||
BC112308.1 | - | 3 | 235 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
