siRNA Details
Gene Symbol: CASC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026939.1 | 5 | 899 | |||
NR_026940.1 | 4 | 799 | ||||
GenBank mRNA | AJ535621.1 | - | 4 | 612 | ||
AY226450.1 | - | 5 | 715 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
