siRNA Details
Gene Symbol: C22orf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182520.2 | NP_872326.2 | 4, 5 | 619 | ||
XM_011529907.2 | XP_011528209.1 | 3, 4 | 1410 | |||
XM_011529908.2 | XP_011528210.2 | 4, 5 | 1131 | |||
XM_011529912.2 | XP_011528214.1 | 2, 3 | 1251 | |||
XM_017028602.1 | XP_016884091.1 | 3 | 1494 | |||
XM_017028603.1 | XP_016884092.1 | 3 | 1473 | |||
XM_017028604.1 | XP_016884093.1 | 4 | 1434 | |||
XM_017028605.1 | XP_016884094.1 | 3, 4 | 1425 | |||
XM_017028606.1 | XP_016884095.1 | 3 | 1494 | |||
XM_017028607.1 | XP_016884096.1 | 3 | 1494 | |||
XM_017028608.1 | XP_016884097.1 | 3 | 1494 | |||
XM_017028609.1 | XP_016884098.1 | 4 | 1223 | |||
XM_017028610.1 | XP_016884099.1 | 4 | 1201 | |||
XM_017028611.1 | XP_016884100.1 | 3, 4 | 1415 | |||
XM_017028612.1 | XP_016884101.1 | 4, 5 | 1115 | |||
XM_017028615.1 | XP_016884104.1 | 4, 5 | 1121 | |||
GenBank mRNA | AK093880.1 | - | 3 | 960 | ||
BC113450.1 | - | 1 | 322 | 8322502 | ||
BC113452.1 | - | 1 | 322 | 8322504 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
