siRNA Details
Gene Symbol: C22orf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182520.2 | NP_872326.2 | 5 | 683 | ||
XM_011529907.2 | XP_011528209.1 | 4 | 1474 | |||
XM_011529908.2 | XP_011528210.2 | 5 | 1195 | |||
XM_011529912.2 | XP_011528214.1 | 3 | 1315 | |||
XM_017028602.1 | XP_016884091.1 | 3 | 1558 | |||
XM_017028603.1 | XP_016884092.1 | 3 | 1537 | |||
XM_017028604.1 | XP_016884093.1 | 4 | 1498 | |||
XM_017028605.1 | XP_016884094.1 | 4 | 1489 | |||
XM_017028606.1 | XP_016884095.1 | 3 | 1558 | |||
XM_017028607.1 | XP_016884096.1 | 3 | 1558 | |||
XM_017028608.1 | XP_016884097.1 | 3 | 1558 | |||
XM_017028609.1 | XP_016884098.1 | 4 | 1287 | |||
XM_017028610.1 | XP_016884099.1 | 4 | 1265 | |||
XM_017028611.1 | XP_016884100.1 | 4 | 1479 | |||
XM_017028612.1 | XP_016884101.1 | 5 | 1179 | |||
XM_017028615.1 | XP_016884104.1 | 5 | 1185 | |||
GenBank mRNA | AK093880.1 | - | 3 | 1024 | ||
BC113450.1 | - | 1 | 386 | 8322502 | ||
BC113452.1 | - | 1 | 386 | 8322504 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
