siRNA Details
Gene Symbol: TMEM97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014573.2 | NP_055388.2 | 3 | 550 | ||
XM_005257965.3 | XP_005258022.1 | 3 | 599 | |||
GenBank mRNA | AK293331.1 | - | 3 | 405 | ||
AK299511.1 | - | 3 | 443 | |||
AK307678.1 | - | 3 | 442 | |||
BC017362.2 | - | 3 | 421 | |||
BC045655.1 | - | 4 | 559 | |||
BC091504.1 | - | 3 | 421 | 30379019 | ||
L19183.1 | - | 3 | 444 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
