siRNA Details
Gene Symbol: MRPS21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018997.3 | NP_061870.1 | 2 | 872 | ||
NM_031901.5 | NP_114107.1 | 3 | 436 | |||
GenBank mRNA | AB049955.1 | - | 3 | 409 | ||
AF182417.1 | - | 2 | 381 | |||
AK058101.1 | - | 2 | 1038 | |||
AK289467.1 | - | 3 | 423 | |||
BC004566.2 | - | 3 | 405 | 3633098 | ||
BC006554.2 | - | 2 | 777 | 2819715 | ||
BI755132.1 | - | 2 | 829 | |||
HY161691.1 | - | 3 | 437 | |||
U79258.1 | - | 2 | 793 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
