siRNA Details
Gene Symbol: DLEC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321153.1 | NP_001308082.1 | 33 | 4609 | ||
NM_007335.3 | NP_031361.2 | 33 | 4600 | |||
NM_007337.3 | NP_031363.2 | 33 | 4600 | |||
XM_006713438.3 | XP_006713501.1 | 33 | 5218 | |||
XM_006713439.3 | XP_006713502.1 | 33 | 5217 | |||
XM_011534317.2 | XP_011532619.1 | 33 | 5217 | |||
XM_011534318.2 | XP_011532620.1 | 32 | 5044 | |||
GenBank mRNA | AB020522.1 | - | 33 | 4600 | ||
AB425198.1 | - | 33 | 4581 | |||
AL137282.1 | - | 2 | 663 | |||
AL137706.1 | - | 21 | 3482 | |||
BC171799.1 | - | 33 | 4600 | 9054453 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
