siRNA Details
Gene Symbol: DLEC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321153.1 | NP_001308082.1 | 34 | 4847 | ||
NM_007335.3 | NP_031361.2 | 34 | 4838 | |||
NM_007337.3 | NP_031363.2 | 34 | 4838 | |||
XM_011534317.2 | XP_011532619.1 | 34 | 5455 | |||
XM_011534318.2 | XP_011532620.1 | 33 | 5282 | |||
GenBank mRNA | AB020522.1 | - | 34 | 4838 | ||
AB425198.1 | - | 34 | 4819 | |||
AL137282.1 | - | 3 | 901 | |||
AL137706.1 | - | 22 | 3720 | |||
BC171799.1 | - | 34 | 4838 | 9054453 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
