siRNA Details
Gene Symbol: ZNF23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304492.1 | NP_001291421.1 | 5 | 890 | ||
NM_001304493.1 | NP_001291422.1 | 7 | 1015 | |||
NM_001304494.1 | NP_001291423.1 | 3 | 608 | |||
NM_145911.2 | NP_666016.1 | 6 | 1462 | |||
GenBank mRNA | AF226996.1 | - | 1 | 40 | ||
AF507946.1 | - | 6 | 1453 | |||
AK297130.1 | - | 3 | 608 | |||
AL080123.1 | - | 1 | 492 | |||
AL833815.1 | - | 2 | 1232 | |||
BC007256.1 | - | 6 | 1453 | 3028040 | ||
BC048974.1 | - | 7 | 1009 | 4816965 | ||
BT007400.1 | - | 3 | 639 | |||
BX537507.1 | - | 1 | 3264 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
