siRNA Details
Gene Symbol: ZNF23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304492.1 | NP_001291421.1 | 5 | 1695 | ||
NM_001304493.1 | NP_001291422.1 | 7 | 1820 | |||
NM_001304494.1 | NP_001291423.1 | 3 | 1413 | |||
NM_145911.2 | NP_666016.1 | 6 | 2267 | |||
GenBank mRNA | AF226996.1 | - | 2 | 761 | ||
AF507946.1 | - | 6 | 2258 | |||
AK091039.1 | - | 5 | 1690 | |||
AK297130.1 | - | 3 | 1413 | |||
AL080123.1 | - | 1 | 1297 | |||
AL833815.1 | - | 2 | 2037 | |||
AY500362.1 | - | 1 | 653 | |||
BC007256.1 | - | 6 | 2258 | 3028040 | ||
BC048974.1 | - | 7 | 1814 | 4816965 | ||
BT007400.1 | - | 3 | 1444 | |||
BX537507.1 | - | 1 | 4069 | |||
X52347.1 | - | 1 | 19 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
