siRNA Details
Gene Symbol: ATXN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000333.3 | NP_000324.1 | 3 | 825 | ||
NM_001177387.1 | NP_001170858.1 | 2 | 382 | |||
GenBank mRNA | AF032105.1 | - | 3 | 825 | ||
AJ000517.1 | - | 3 | 833 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
