siRNA Details
Gene Symbol: SNX21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042632.2 | NP_001036097.1 | 4 | 618 | ||
NM_001042633.2 | NP_001036098.1 | 5 | 636 | |||
NM_033421.3 | NP_219489.1 | 4 | 625 | |||
NM_152897.2 | NP_690857.1 | 4 | 625 | |||
XM_005260608.4 | XP_005260665.1 | 2 | 700 | |||
XM_011529095.2 | XP_011527397.1 | 2 | 604 | |||
GenBank mRNA | AF258570.1 | - | 1 | 285 | ||
AF395845.1 | - | 4 | 536 | |||
AF523834.1 | - | 4 | 604 | |||
AK091359.1 | - | 3 | 640 | |||
AK095851.1 | - | 3 | 499 | |||
AK226120.1 | - | 3 | 149 | |||
AK293443.1 | - | 5 | 637 | |||
AK297622.1 | - | 4 | 559 | |||
AL591562.1 | - | 4 | 604 | |||
BC012904.1 | - | 6 | 856 | |||
BC019823.1 | - | 4 | 598 | 4453502 | ||
BM925584.1 | - | 5 | 622 | |||
KU178820.1 | - | 4 | 536 | |||
KU178821.1 | - | 5 | 547 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
