siRNA Details
Gene Symbol: SNX21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042632.2 | NP_001036097.1 | 4 | 543 | ||
NM_001042633.2 | NP_001036098.1 | 5 | 561 | |||
NM_033421.3 | NP_219489.1 | 4 | 550 | |||
NM_152897.2 | NP_690857.1 | 4 | 550 | |||
XM_005260608.4 | XP_005260665.1 | 2 | 625 | |||
XM_011529095.2 | XP_011527397.1 | 2 | 529 | |||
XM_011529098.2 | XP_011527400.1 | 5 | 1063 | |||
GenBank mRNA | AF395845.1 | - | 4 | 461 | ||
AF523834.1 | - | 4 | 529 | |||
AK091359.1 | - | 3 | 565 | |||
AK095851.1 | - | 3 | 424 | |||
AK297622.1 | - | 4 | 484 | |||
AL591562.1 | - | 4 | 529 | |||
BC012904.1 | - | 6 | 781 | |||
BM925584.1 | - | 5 | 547 | |||
KU178820.1 | - | 4 | 461 | |||
KU178821.1 | - | 5 | 472 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
