siRNA Details
Gene Symbol: SNX21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042633.2 | NP_001036098.1 | 3 | 522 | ||
NM_033421.3 | NP_219489.1 | 3 | 522 | |||
NM_152897.2 | NP_690857.1 | 3 | 522 | |||
XM_011529098.2 | XP_011527400.1 | 3 | 935 | |||
XM_011529099.2 | XP_011527401.1 | 3 | 935 | |||
XM_017028120.1 | XP_016883609.1 | 3 | 935 | |||
GenBank mRNA | AF395845.1 | - | 3 | 433 | ||
AF523834.1 | - | 3 | 501 | |||
AK091359.1 | - | 1 | 526 | |||
AK226120.1 | - | 1 | 35 | |||
AK293443.1 | - | 3 | 523 | |||
AK297622.1 | - | 2 | 445 | |||
AL591562.1 | - | 3 | 501 | |||
BC012904.1 | - | 3 | 522 | |||
BC019823.1 | - | 3 | 495 | 4453502 | ||
BM925584.1 | - | 3 | 508 | |||
DA053179.1 | - | 3 | 523 | |||
KU178820.1 | - | 3 | 433 | |||
KU178821.1 | - | 3 | 433 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
