siRNA Details
Gene Symbol: SYMPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004819.2 | NP_004810.2 | 19 | 2814 | ||
XM_005259286.1 | XP_005259343.1 | 19 | 2767 | |||
XM_011527354.1 | XP_011525656.1 | 20 | 2817 | |||
GenBank mRNA | AB209024.1 | - | 19 | 4977 | ||
AB210010.1 | - | 19 | 2659 | |||
AK130602.1 | - | 10 | 953 | |||
BC006536.2 | - | 18 | 2521 | |||
BC006567.2 | - | 18 | 2521 | |||
BX649165.1 | - | 18 | 3720 | |||
U49240.1 | - | 14 | 2283 | |||
Y10931.1 | - | 19 | 2632 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
