siRNA Details
Gene Symbol: SYMPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004819.2 | NP_004810.2 | 22 | 3106 | ||
XM_005259286.1 | XP_005259343.1 | 22 | 3059 | |||
XM_011527354.1 | XP_011525656.1 | 23 | 3109 | |||
GenBank mRNA | AB209024.1 | - | 22 | 5269 | ||
AB210010.1 | - | 22 | 2951 | |||
AK130602.1 | - | 13 | 1245 | |||
AK300252.1 | - | 22 | 2960 | |||
BC006536.2 | - | 21 | 2813 | |||
BC006567.2 | - | 21 | 2813 | |||
BX649165.1 | - | 21 | 4012 | |||
U49240.1 | - | 17 | 2575 | |||
Y10931.1 | - | 22 | 2924 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
