siRNA Details
Gene Symbol: SYMPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004819.2 | NP_004810.2 | 17 | 2544 | ||
XM_005259286.1 | XP_005259343.1 | 17 | 2497 | |||
XM_011527354.1 | XP_011525656.1 | 18 | 2547 | |||
GenBank mRNA | AB209024.1 | - | 17 | 4707 | ||
AB210010.1 | - | 17 | 2389 | |||
AK130602.1 | - | 8 | 683 | |||
AK300252.1 | - | 17 | 2398 | |||
BC006536.2 | - | 16 | 2251 | |||
BC006567.2 | - | 16 | 2251 | |||
BX649165.1 | - | 16 | 3450 | |||
U49240.1 | - | 12 | 2013 | |||
Y10931.1 | - | 17 | 2362 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
