siRNA Details
Gene Symbol: ANKRD33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130015.1 | NP_001123487.1 | 6 | 1496 | ||
NM_001304459.1 | NP_001291388.1 | 6 | 1759 | |||
NM_001304460.1 | NP_001291389.1 | 5 | 1370 | |||
NM_182608.3 | NP_872414.3 | 5 | 1670 | |||
GenBank mRNA | AF517120.1 | - | 1 | 253 | ||
AK125517.1 | - | 5 | 1719 | |||
BX537908.1 | - | 6 | 1271 | |||
BX640656.1 | - | 6 | 1271 | |||
BX647684.1 | - | 2 | 2970 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
