siRNA Details
Gene Symbol: PARD6G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032510.3 | NP_115899.1 | 3 | 864 | ||
GenBank mRNA | AB043635.2 | - | 1 | 403 | ||
AB044556.1 | - | 3 | 698 | |||
BC060797.1 | - | 3 | 864 | 30348280 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
