siRNA Details
Gene Symbol: HOXA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005522.4 | NP_005513.1 | 1 | 330 | ||
NM_153620.2 | NP_705873.2 | 1 | 330 | |||
GenBank mRNA | AK308067.1 | - | 1 | 278 | ||
AK313514.1 | - | 1 | 321 | |||
BC032547.2 | - | 1 | 298 | 5537563 | ||
DB246535.1 | - | 1 | 330 | |||
S79869.1 | - | 1 | 230 | |||
S79871.1 | - | 1 | 230 | |||
S79910.1 | - | 1 | 230 | |||
U10421.1 | - | 1 | 236 | |||
U37431.1 | - | 1 | 230 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
