siRNA Details
Gene Symbol: HOXA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005522.4 | NP_005513.1 | 1 | 435 | ||
NM_153620.2 | NP_705873.2 | 1 | 435 | |||
GenBank mRNA | AK308067.1 | - | 1 | 383 | ||
AK313514.1 | - | 1 | 426 | |||
BC032547.2 | - | 1 | 403 | 5537563 | ||
DB246535.1 | - | 1 | 435 | |||
S79869.1 | - | 1 | 335 | |||
S79871.1 | - | 1 | 335 | |||
S79910.1 | - | 1 | 335 | |||
U10421.1 | - | 1 | 341 | |||
U37431.1 | - | 1 | 335 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
