siRNA Details
Gene Symbol: RIC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135109.2 | NP_001128581.1 | 2 | 235 | ||
NM_001206671.2 | NP_001193600.1 | 6 | 781 | |||
NM_001206672.2 | NP_001193601.1 | 4 | 538 | |||
NM_024557.4 | NP_078833.3 | 6 | 778 | |||
NR_045405.1 | 7 | 830 | ||||
XM_006718317.3 | XP_006718380.1 | 6 | 910 | |||
XM_006718318.3 | XP_006718381.1 | 6 | 823 | |||
XM_017018287.1 | XP_016873776.1 | 6 | 910 | |||
XM_017018288.1 | XP_016873777.1 | 7 | 907 | |||
XM_017018289.1 | XP_016873778.1 | 7 | 854 | |||
XM_017018290.1 | XP_016873779.1 | 5 | 677 | |||
GenBank mRNA | AK021670.1 | - | 5 | 802 | ||
AK293933.1 | - | 6 | 828 | |||
AK296505.1 | - | 7 | 795 | |||
AK315379.1 | - | 6 | 737 | |||
AL832601.2 | - | 2 | 196 | |||
AM422214.1 | - | 4 | 484 | |||
AY326435.1 | - | 6 | 727 | |||
BC022455.1 | - | 6 | 760 | 4792930 | ||
BX647514.1 | - | 3 | 378 | |||
DA186330.1 | - | 2 | 235 | |||
KU178773.1 | - | 6 | 724 | |||
KU178774.1 | - | 5 | 630 | |||
KU178775.1 | - | 2 | 181 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
