siRNA Details
Gene Symbol: RIC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135109.2 | NP_001128581.1 | 2 | 809 | ||
NM_001206671.2 | NP_001193600.1 | 6 | 1355 | |||
NM_001206672.2 | NP_001193601.1 | 4 | 1112 | |||
NM_024557.4 | NP_078833.3 | 6 | 1352 | |||
NR_045405.1 | 7 | 1404 | ||||
XM_017018287.1 | XP_016873776.1 | 6 | 1484 | |||
XM_017018288.1 | XP_016873777.1 | 7 | 1481 | |||
XM_017018289.1 | XP_016873778.1 | 7 | 1428 | |||
XM_017018290.1 | XP_016873779.1 | 5 | 1251 | |||
GenBank mRNA | AK021670.1 | - | 5 | 1376 | ||
AL832601.2 | - | 2 | 770 | |||
AY326435.1 | - | 6 | 1301 | |||
BC022455.1 | - | 6 | 1334 | 4792930 | ||
BX647514.1 | - | 3 | 952 | |||
DQ347704.1 | - | 1 | 98 | |||
DQ347705.1 | - | 1 | 97 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
