siRNA Details
Gene Symbol: RIC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135109.2 | NP_001128581.1 | 2 | 411 | ||
NM_001206671.2 | NP_001193600.1 | 6 | 957 | |||
NM_001206672.2 | NP_001193601.1 | 4 | 714 | |||
NM_024557.4 | NP_078833.3 | 6 | 954 | |||
NR_045405.1 | 7 | 1006 | ||||
XM_006718317.3 | XP_006718380.1 | 6 | 1086 | |||
XM_006718318.3 | XP_006718381.1 | 6 | 999 | |||
XM_017018287.1 | XP_016873776.1 | 6 | 1086 | |||
XM_017018288.1 | XP_016873777.1 | 7 | 1083 | |||
XM_017018289.1 | XP_016873778.1 | 7 | 1030 | |||
XM_017018290.1 | XP_016873779.1 | 5 | 853 | |||
GenBank mRNA | AK021670.1 | - | 5 | 978 | ||
AK296505.1 | - | 7 | 971 | |||
AK315379.1 | - | 6 | 913 | |||
AL832601.2 | - | 2 | 372 | |||
AM422214.1 | - | 4 | 660 | |||
AY326435.1 | - | 6 | 903 | |||
BC022455.1 | - | 6 | 936 | 4792930 | ||
BX647514.1 | - | 3 | 554 | |||
DA186330.1 | - | 2 | 411 | |||
KU178773.1 | - | 6 | 900 | |||
KU178774.1 | - | 5 | 806 | |||
KU178775.1 | - | 2 | 357 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
