siRNA Details
Gene Symbol: ZDHHC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024786.2 | NP_079062.1 | 13 | 1900 | ||
XM_017009869.1 | XP_016865358.1 | 14 | 2057 | |||
XM_017009870.1 | XP_016865359.1 | 16 | 3868 | |||
XM_017009871.1 | XP_016865360.1 | 16 | 3817 | |||
XM_017009872.1 | XP_016865361.1 | 14 | 1892 | |||
XM_017009873.1 | XP_016865362.1 | 10 | 1874 | |||
XM_017009874.1 | XP_016865363.1 | 10 | 2754 | |||
XM_017009875.1 | XP_016865364.1 | 10 | 2701 | |||
XM_017009876.1 | XP_016865365.1 | 10 | 1668 | |||
XM_017009877.1 | XP_016865366.1 | 10 | 1242 | |||
XM_017009878.1 | XP_016865367.1 | 10 | 1199 | |||
GenBank mRNA | AF267859.1 | - | 2 | 316 | ||
AK023215.1 | - | 13 | 1900 | |||
AK024233.1 | - | 12 | 3218 | |||
AK096220.1 | - | 1089 | ||||
BC032000.1 | - | 13 | 1519 | 4750082 |
Gene Symbol: ZDHHC11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017010116.1 | XP_016865605.1 | 14 | 3497 | ||
XM_017010120.1 | XP_016865609.1 | 15 | 4445 | |||
XM_017010122.1 | XP_016865611.1 | 15 | 4442 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
