siRNA Details
Gene Symbol: RPS21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024.3 | NP_001015.1 | 4 | 238 | ||
GenBank mRNA | AV758927.1 | - | 199 | |||
BC018140.1 | - | 3 | 175 | 3903320 | ||
BC021990.1 | - | 4 | 203 | 4066330 | ||
BM840443.1 | - | 4 | 238 | |||
CF124568.1 | - | 4 | 279 | |||
CR542132.1 | - | 3 | 148 | |||
L04483.1 | - | 4 | 186 | |||
X79563.2 | - | 4 | 184 |
Gene Symbol: TAPT1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AI770046.1 | - | 1 | 570 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
