siRNA Details
Gene Symbol: NXPE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134456.1 | NP_001127928.1 | 7 | 1919 | ||
NM_145037.2 | NP_659474.1 | 7 | 1600 | |||
GenBank mRNA | AF173890.1 | - | 3 | 246 | ||
AK054664.1 | - | 4 | 1023 | |||
AK289689.1 | - | 7 | 1919 | |||
AY358207.1 | - | 6 | 1864 | |||
BC009431.1 | - | 7 | 1555 | 3163718 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
