siRNA Details
Gene Symbol: IGFLR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024660.2 | NP_078936.1 | 5 | 871 | ||
GenBank mRNA | AK026226.1 | - | 5 | 872 | ||
AX775837.1 | - | 5 | 874 | |||
BC031329.1 | - | 2 | 364 | |||
BC126481.1 | - | 5 | 832 | 8992197 | ||
BC126483.1 | - | 5 | 832 | 8992199 | ||
BC144205.1 | - | 5 | 830 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
